Rubbermaid® Tilt Trucks
Leverancier: Rubbermaid Commercial Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FG9T1500BLA FG101300BLA FG9T1400BLA FG9T1600BLA FG9T1300BLA FG101100BLA FG102800BLA FG9T2200BLA FG9T2300BLA
129-1966EA 2130 EUR
129-1966 129-2117 129-1967 129-1965 129-1968 129-2166 129-2205 129-1964 129-1950
Rubbermaid® Tilt Trucks
Transportwagens
The Rubbermaid commercial tilt truck, structural foam, offers industrial strength construction to transport heavy loads up to 955 kg (with 0,8 m³ heavy-duty model).
  • Flexible movement - These dump trucks fit through standard commercial doorways and onto elevators easily
  • The constant force design allows for controlled dumping by one person
  • Ergonomic 'steering wheel' design handle enhances maneuverability
  • Recycled material - Contains up to 70% post consumer recycled resin (PCR) exceeding EPA guidelines
  • Easy to clean - maintenance-free
  • Unibody design - Promotes a more discreet appearance in public areas
  • Leakproof body
The non-marking casters reduce touch-up cleaning.
Bestelinformatie: Optional domed lid is available.