Polycarbin® Racked LTS® Compatible Pipette Tips
Leverancier: Polycarbin
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC01-04-05 PC01-04-06 PC01-05-05 PC01-05-06 PC01-06-05 PC01-06-06
732-3985EA 1110 EUR
732-3985 613-7227 732-3986 613-7228 732-3987 613-7229
Polycarbin® Racked LTS® Compatible Pipette Tips
Pipettips
Polycarbin’s pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint (tips do not contain recycled content).
  • Rainin LTS® compatible
  • Carries a smaller carbon footprint
  • DNase, RNase and pyrogen free
  • Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, we focused on the the tip box, or 'rack,' which is the most dense, carbon-intensive piece of plastic. All tip boxes, of any brand, can be remanufactured and transformed into the next line of lab products using Polycarbin's alpha carbin program, a mailback service (sold separately).
Certificaten: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Verpakking: Pipette tips are sold by the case. Each case contains 5 packs of 10 racks (50 racks total, 4800 tips).