Sensitivity Solutions
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FT11 FT-31
MMMAFT11EA 277 EUR
MMMAFT11 MMMAFT-31
Sensitivity Solutions
Ademhalingsmaskers
3M™ sensitivity solutions are used to test the sensitivity of the respirator wearer to a sweet or bitter solution. These solutions are intended for use with 3M™ qualitative fit test apparatus kits FT-10 (sweet) and FT-30 (bitter).
  • Sweet and bitter taste sensitivity solutions for fit testing
  • 55 ml bottle
  • For occupational use only
Test kits and accessories designed to help organisations meet regulatory fit testing requirements for respirators. Sensitivity solutions are used to identify whether the test recipient can detect the solution prior to wearingMore Product Information