HTD Equilibrium Dialysis Device
Leverancier: HTDIALYSIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1103 1006 1104 1105 1009 1101 1135 1151
HTDL1103EA 0 EUR
HTDL1103 HTDL1006 HTDL1104 HTDL1105 HTDL1009 HTDL1101 HTDL1135 HTDL1151
HTD Equilibrium Dialysis Device
Slangen Dialyseslangen
HTD reusable equilibrium dialysis device combines industry-leading accuracy and precision with an automation compatible design for a cost efficient, scalable solution.
  • Simultaneously process 1 to 96 assays, each with a 30 to 150 µl capacity to conserve valuable reagents
  • Compatible with a wide range of dialysis membrane pore sizes
  • Ready in minutes, achieves equilibrium in 4 to 6 hours on average
  • Automatable with standard robotic workstations and 96-well pipettors
  • Ready access to both the sample and dialysate sides of the membrane from the top of the device
  • Minimal non-specific binding due to 100% Teflon block construction
  • Reusable device is cost-effective, eco-friendly, and scalable
  • Compatible with a wide range of dialysis membrane pore sizes: 1 kDa, 3,5 kDa, 6 to 8 kDa, 10 kDa, 12 to 14 kDa, 25 kDa, 50 kDa
The HTD96B micro equilibrium device was developed at a Global Pharmaceutical Research laboratory to assess serum protein binding, microsomal protein binding, and tissue specific binding of drug candidates, conserving sample quantities, and achieving rapid equilibrium.
Reusable device combines industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solution.
Applications:
- Serum/plasma protein binding
- Microsome protein binding
- Relative binding of drug to specific tissues
- Receptor binding
- T3/T4 free fraction binding (hyperthyroidism) determinations
The 96C model has a shorter diffusion path with a 25 to 75 µl capacity and achieves equilibrium in 2 to 4 hours on average.
Additional specifications:
Dimensions: 170,1×116,8×38,1 mm
Weight: 0,54 kg