Gel-clot Reaction Tubes
Leverancier: FUJIFILM WAKO CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CT-1075 CT-1075B
WAKFCT-1075EA 125 EUR
WAKFCT-1075 WAKFCT-1075B
Gel-clot Reaction Tubes
Buffers Biologische buffers
These Gel-clot Reaction Tubes are made of high-quality borosilicate glass, depyrogenated at 250 °C. This results in an endotoxin-free tube that is designed to perform successful LAL gel clot assays with heat blocks and water baths.
  • Endotoxin-free test tubes (<0,001 EU/tube)
  • Endotoxin-free tubes and caps sterilised at 250 °C
  • Made with borosilicate glass and designed for gel clot testing