Vis-Tex Cut Gloves Long Cuff
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A631O8RXL A631O8RXXL A631O8RXXXL A631O8RL A631O8RM A631O8RS
PRTWA631O8RXLEA 84 EUR
PRTWA631O8RXL PRTWA631O8RXXL PRTWA631O8RXXXL PRTWA631O8RL PRTWA631O8RM PRTWA631O8RS
Vis-Tex Cut Gloves Long Cuff
Handschoenen
Cut level D glove with PU dipping for high abrasion resistance, the HPPE and Glass Fibre liner also achieves a level 1 for contact heat.
  • Level D cut resistance
  • 13 gauge liner for a perfect fit
  • Smooth PU coating for increased abrasion resistance
  • Extended cuff for wrist and forearm protection
  • High visibility liner
  • Tested for both cut and heat protection
  • Palm dipped to increase dexterity and ventilation
  • Breathable seamless liner
  • ANSI cut level A4
Flexible hi-vis lining for optimum protection and comfort. Extended 10 cm cuff adds additional protection to the wrist and forearm area. Retail tag aids presentation for retail sales.
Colours: Orange/black
Certificaten: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4X43D, EN 407: 2020 X1XXXX. ANSI/ISEA 105: 2016 CUT Level A4, ANSI/ISEA 105: 2016 ABRASION Level 6. CE certified.