Antistatic Shell Gloves, A197
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A197GRRL A197GRRM A197GRRS A197GRRXL A197GRRXS
PRTWA197GRRLEA 14.6 EUR
PRTWA197GRRL PRTWA197GRRM PRTWA197GRRS PRTWA197GRRXL PRTWA197GRRXS
Antistatic Shell Gloves, A197
Handschoenen
Constructed from an advanced polyester and carbon fibre blend. This glove protects equipment like circuit boards from the natural oils of the skin while also protecting sensitive equipment from harmful static discharge.
  • Suitable for use in ESD environments
  • This glove can be used with most mobile touchscreen devices
  • Seamless 13 gauge liner
Low linting construction for minimal contamination. Retail tag aids presentation for retail sales.
Colour: Grey
Certificaten: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 113XX, EN 16350:2014 Pass Level R <1.0 x 108 Ω -. IEC 61340-5-1:2016. CE certified.