CS Cut F13 PU Gloves
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A670K8RL A670K8RM A670K8RS A670K8RXL A670K8RXS A670K8RXXL
PRTWA670K8RLEA 109 EUR
PRTWA670K8RL PRTWA670K8RM PRTWA670K8RS PRTWA670K8RXL PRTWA670K8RXS PRTWA670K8RXXL
CS Cut F13 PU Gloves
Handschoenen
Suitable for use with most touchscreen devices, this glove features a reinforced thumb crotch for extra durability.
  • Level F cut resistance
  • Protection against contact heat up to 100 °C
  • Smooth PU coating for increased abrasion resistance
  • Reflective label for improved visibility
  • Seamless 13 gauge liner
  • Palm dipped to increase dexterity and ventilation
  • ANSI cut level A6
The breathable 13-gauge liner ensures an excellent fit and dexterity. PU Palm coating offers secure grip in dry and light oily handling environments. Reflective label increases glove visibility in low light conditions. Retail tag aids presentation for retail sales.
Colour: Black
Certificaten: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4X43F, EN 407: 2020 X1XXXX. ANSI/ISEA 105: 2016 CUT Level A6, ANSI/ISEA 105: 2016 ABRASION Level 6, ANSI/ISEA 105: 2016 PUNCTURE Level 3. CE certified.