Eco Hi-Vis Winter Jackets
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EC60ORR4XL EC60ORRL EC60ORRM EC60ORRS EC60ORRXL EC60ORRXXL EC60ORRXXXL EC60YER4XL EC60YERL EC60YERM EC60YERS EC60YERXL EC60YERXXL EC60YERXXXL
PRTWEC60ORR4XLEA 73.3 EUR
PRTWEC60ORR4XL PRTWEC60ORRL PRTWEC60ORRM PRTWEC60ORRS PRTWEC60ORRXL PRTWEC60ORRXXL PRTWEC60ORRXXXL PRTWEC60YER4XL PRTWEC60YERL PRTWEC60YERM PRTWEC60YERS PRTWEC60YERXL PRTWEC60YERXXL PRTWEC60YERXXXL
Eco Hi-Vis Winter Jackets
Werkkleding
This jacket offers exceptional weather protection, comfort, and warmth while utilising the latest innovations in recycled fabrics. The outer shell, lining, and wadding are certified recycled polyester, and the packaging is derived from sugarcane, allowing the wearer to champion sustainability without compromising on performance.
  • Eco-conscious certified recycled fabric
  • Waterproof with taped seams preventing water penetration
  • Quilt lined for thermal insulation
  • Articulated darts for additional shaping
  • Made using recycled polyester
  • Print access for corporate branding
  • 40 UPF rated fabric to block 98% of UV rays
  • Concealed mobile phone pocket
  • Certified to EN ISO 20471 after 50x washes
  • Detachable hood
  • Reflective tape for increased visibility
The Eco Hi-Vis winter jacket is manufactured from an extremely water-resistant eco-conscious fabric finish, allowing water to bead away from the fabric surface. Jacket features include an internal chest pocket, drawcord adjustable hem, hook and loop cuffs for a secure fit, and detachable lined hood. A radio loop is included for easy clipping of a radio, as well as 4 pockets for ample storage, a storm flap front to protect against the elements, and a chin guard for added comfort and stability.
Colour: Yellow or orange
Weather conditions: Dark: High visibility, reflective clothing
Certificaten: EN ISO 20471 Class 3, EN 343 Class 3:1 X WP 15,000mm. RIS 3279 TOM Issue 2 (Orange only). ANSI/ISEA 107 Type R Class 3, ANSI/ISEA 107 Type P Class 3. CE certified.