Safety Ankle Boots, Lace-Up, High, Compositelite All Weather Boot S3 WR
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FC57BKR38 FC57BKR39 FC57BKR40 FC57BKR41 FC57BKR42 FC57BKR43 FC57BKR44 FC57BKR45 FC57BKR46 FC57BKR47 FC57BKR48
PRTWFC57BKR38EA 136 EUR
PRTWFC57BKR38 PRTWFC57BKR39 PRTWFC57BKR40 PRTWFC57BKR41 PRTWFC57BKR42 PRTWFC57BKR43 PRTWFC57BKR44 PRTWFC57BKR45 PRTWFC57BKR46 PRTWFC57BKR47 PRTWFC57BKR48
Safety Ankle Boots, Lace-Up, High, Compositelite All Weather Boot S3 WR
Schoenen en laarzen
These 100% non-metallic boots offers superb lightweight protection for feet, even in wet conditions with a waterproof and breathable lining. Dual density PU/TPU hardwearing outsole with shock absorbing seat region, antistatic and slip-resistant properties.
  • Waterproof membrane for dry foot comfort
  • Angled top line for better fit and leg flexibility
  • Protective scuff cap extends longevity of the boot
  • Textile areas aid ventilation and airflow to the foot
  • Full grain leather
  • Fuel- and oil-resistant outsole
  • Dual density sole unit; wide fitting
The Compositelite all weather boot features a padded collar for added comfort, composite toecap for added protection, pierce-resistant composite midsole, and a protective scuff cap. SRC - slip resistant outsole to prevent slips and trips on ceramic and steel surfaces.
Colour: Black
Certificaten: EN ISO 20345: 2011 S3. CE certified.