SymLine Lab Sinks with Drain and Mounting Sleeve
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
118020 118021 118024 118025
SCTV118020EA 580 EUR
SCTV118020 SCTV118021 SCTV118024 SCTV118025
SymLine Lab Sinks with Drain and Mounting Sleeve
Kranen en gootstenen Gootstenen
For integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.
  • Made of dissipative PE-HD-EL
  • Base with integrated slope for complete emptying
  • Drain with removable sieve
  • Laterally mounted earthing cable (length 1,5 m)
Secure mounting in the worktop is ensured by the fastening sleeves.
Dimension: 401×300×190 mm