Eyewear cleaning station, uvex
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9970.005 9973.101 9972.103
UVEX9970.005EA 77.8 EUR
UVEX9970.005 UVEX9973.101 UVEX9972.103
Eyewear cleaning station, uvex
Reinigingsmiddelen Lens Cleaning Solutions
This lens cleaning station is wall-mountable and therefore ensures a quick and easy access for your employees.
  • Suitable for all uvex safety spectacles
  • Easy refilling, spare parts can be ordered separately
  • Slim and robust design, compact size (24 × 28 cm)
  • Closed rear cover to prevent contamination of the cleaning accessories
  • Easy to handle with 1-click operation
  • Easy to mount on walls (with screws)
The station comes equipped with one bottle of cleaning fluid (0,5 L), a plastic pump that can be attached to the bottle of cleaning fluid, and anti-static, silicone-free lens cleaning papers (approx. 700 sheets).