Hi-Vis Women's Breathable Rain Jackets
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LW70YERL LW70YERM LW70YERS LW70YERXL LW70YERXS LW70YERXXL LW70YERXXXL
PRTWLW70YERLEA 60.3 EUR
PRTWLW70YERL PRTWLW70YERM PRTWLW70YERS PRTWLW70YERXL PRTWLW70YERXS PRTWLW70YERXXL PRTWLW70YERXXXL
Hi-Vis Women's Breathable Rain Jackets
Werkkleding
An extremely stylish women's classic hi-vis jacket. The contemporary silhouette together with the angled reflective tape ensures an attractive and flattering fit.
  • Durable, breathable, windproof and water-resistant fabric
  • Zipped pockets
  • Hook and loop cuffs for a secure fit
  • Curved back hem for added protection
  • Concealed mobile phone pocket
  • Four pockets for ample storage
  • 40+ UPF rated fabric to block 98% of UV rays
The breathable and durable oxford fabric, curved back hem, detachable hood, and secure zipped pockets makes this jacket suitable for multiple working environments. This garment features an extremely water-resistant, eco-conscious fabric finish that allows water to bead away from the fabric surface. Taped seams further prevent water penetration. Additional features include a storm flap front to protect against the elements, reflective tape for increased visibility, raglan sleeves for a comfort fit, and a pack-away detachable hood for convenience.
Certificaten: EN ISO 20471 Class 3 after 50× washes. RIS 3279 TOM Issue 2 (orange only). EN 343 Class 4:3 X WP 10000 mm, MVP 10000 g/m²/24 h. ANSI/ISEA 107 Type R Class 3. ANSI/ISEA 107 Type P Class 3.