VWR® Vial Grippers
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-2990EA 62.8 EUR
216-2990 216-2991 216-2989
VWR® Vial Grippers
Pincetten
These two styles of hand-operated grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively.
  • Each style is available in a pack of five assorted colours (red, blue, green, purple, yellow) for easy identification and organisation
  • A combo pack with one of each style is also available
  • Reinforced polypropylene
  • Autoclavable
  • Reusable
  • Plastic-free packaging
Both are durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear. The designated squeeze target ensures maximum precision and grip with every use.