Colour Polyester Labels for M510, BMP51, BMP53 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170906 170925 170902 170903 170904 170905 170907 170908 170909 170910 170911 170912 170913 170914 170915 170916 170917 170918 170919 170920 170921 170922 170923 170924
BRDY170906EA 59.06 EUR
BRDY170906 BRDY170925 BRDY170902 BRDY170903 BRDY170904 BRDY170905 BRDY170907 BRDY170908 BRDY170909 BRDY170910 BRDY170911 BRDY170912 BRDY170913 BRDY170914 BRDY170915 BRDY170916 BRDY170917 BRDY170918 BRDY170919 BRDY170920 BRDY170921 BRDY170922 BRDY170923 BRDY170924
Colour Polyester Labels for M510, BMP51, BMP53 and M511 Printers
Etiketten
Easily differentiate lab samples with the B-494 colour polyester labels, available in six colour options. Pair with a versatile Brady portable label printer and print almost anywhere.
  • Ideal for easily differentiating vials, PCR and microcentrifuge tubes, and slides
  • Designed to withstand abrasions and exposure to chemicals and extreme temperatures, including immersion in liquid nitrogen and hot water baths
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces such as glass and polypropylene
  • Gloss finish
Perfect for cold temperature labelling, component and equipment labelling, and laboratory labelling applications. Gloss finish, chemical-resistant, smear-resistant, high and low temperature-resistant, indoor use B-494 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511. They should be used with the ribbon series R6400.
Available in six colour options with a colour bar across the top of the label; pre-sized for multiple applications, some options with matching colour vial top label.
Certificaten: RoHS compliant to 2011/65/EU (Pass 2015/863).