Safety Storage Cabinet, Type 90, ION-PRO-90 CHARGE model IO90.195.120.PC.WDC
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41269047 41391 41392 41393 41394
135-2884EA 6910 EUR
135-2884 MSGA41391 MSGA41392 MSGA41393 MSGA41394
Safety Storage Cabinet, Type 90, ION-PRO-90 CHARGE model IO90.195.120.PC.WDC
Meubilair Opslagkasten
Safe active and passive storage of lithium-ion batteries.
  • Robust construction and longevity; Convenient cabinet entry
  • Integrated potential-free alarm contact; No unauthorised use
  • Integrated transport base; Easy alignment
  • Smoke detector and temperature sensor; Extraction unit
  • Pressure relief; All-around protection
  • LED display
  • Fire suppression system
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Convenient cabinet entry: open both doors at the same time with just one hand. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with relay module and module for remote signalling.
No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green). Permanent locking of the doors by automatic 2-point locking. Integrated transport base: In the event of an emergency, immediate evacuation and transport of the cabinet to a safe place outdoors.
Easy alignment: adjusting aids to compensate for uneven floor. Smoke detector and temperature sensor for fire detection and forwarding the alarm to the building management system. Extraction unit to prevent heat build-up during battery charging. Pressure relief in the head section.
All-around protection: 90 minutes fire protection from outside to inside (type 90/type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside. LED display with visual and audible alarm for a quick indication of emergencies without opening the cabinet doors. Fire suppression system: Triggers in the event of a fire inside the cabinet automatically.
Certificaten: Fire resistance 90 minutes. Type tested in accordance with EN 14470-1. CE compliant.
Informatie over toebehoren: The optional relay module (sold separately) can differentiate between 4 different warning and alarm levels (Warning, Alarm level 1, Alarm level 2 and power failure) and passes them on to a control center via separate potential-free contacts either as make contacts or break contacts 1× connector for customer wiring for one of the 4 warning and alarm levels is included in the delivery.
Note: The switching contacts are used to connect a signal to a building management system. Direct integration into a fire alarm control panel (FACP) is not recommended or may only be carried out in consultation with the person responsible for the system.
Bestelinformatie: Interior equipment packages need to be ordered separately (not included). Please contact your Avantor representative for more information and available options.
Leveringsinformatie: Delivery prices available upon request.
Opgelet: According to the security concept it is highly recommended to place ION-LINE cabinets at the ground level in a location from where a quick evacuation from the building is possible in the event of fire.