VisTex SMS Coveralls, Type 5/6
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST36ORRL ST36ORRM ST36ORRXL ST36ORRXXL ST36ORRXXXL
PRTWST36ORRLEA 413 EUR
PRTWST36ORRL PRTWST36ORRM PRTWST36ORRXL PRTWST36ORRXXL PRTWST36ORRXXXL
VisTex SMS Coveralls, Type 5/6
Werkkleding
This SMS coverall features Hi-Vis reflective tape for enhanced visibility at night or in dim lighting conditions. This trilaminate constructed fabric offers a high tensile strength and toughness. It provides a fluid and particulate barrier to protect the worker without hindering mobility and comfort.
  • Reflective tape for increased visibility
  • Provides an effective barrier against particulates, aerosols and light sprays
  • Anti-static
  • Designed with a comfort fit
  • Elasticated cuffs for a secure fit
  • Ergonomically shaped hood for a perfect fit
  • External standards label for quick identification of protection levels
  • Two-way zip for quick and easy access
  • Retail bag which aids presentation for retail sales
  • CE-CAT III
Colour: Orange
Weather conditions: Dark - high visibility, reflective clothing
Certificaten: EN ISO 13982 Type 5; EN 13034 Type 6; EN 1073-2; EN 1149 -5.