FreezerBondz™ Cryogenic Matte Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170890 170775 170776 170888 170889 170891
BRDY170890EA 84.74 EUR
BRDY170890 BRDY170775 BRDY170776 BRDY170888 BRDY170889 BRDY170891
FreezerBondz™ Cryogenic Matte Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Securely mark flat laboratory surfaces in liquid nitrogen or freezer environments with the durable B-490 polyester label material. Easily print almost anywhere with a Brady handheld label printer.
  • For laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws, and slides
  • Withstands solvents, smudges, and extreme temperatures
  • Made with a permanent acrylic adhesive
These specialised labels are designed for laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws, and slides. The durable construction withstands solvents, smudges, and extreme temperatures, including storage in liquid nitrogen. The permanent acrylic adhesive can be used on flat frozen surfaces or for wrapping around tubes and vials with a 1/8" overlap. For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
Matte finish, chemical-resistant, smear-resistant, UV-resistant, weather-resistant B-490 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certificaten: RoHS compliant to 2011/65/EU (Pass 2015/863).
Verpakking: Labels are packaged 180 per box.