High-voltage gloves, ActivArmr® 96-003
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
96-003/10 96-003/11 96-003/12 96-003/8 96-003/9
ANSE96-003/10EA 454 EUR
ANSE96-003/10 ANSE96-003/11 ANSE96-003/12 ANSE96-003/8 ANSE96-003/9
High-voltage gloves, ActivArmr® 96-003
Handschoenen
Goatskin leather protectors, for extended high-voltage glove life and enhanced hand safety.
  • Straight cuff with overedge binding fabric
  • Robust goatskin leather: Puncture, abrasion and cut resistance
  • Protective design: For longer-lasting electrical safety gloves
  • Leather composition enhances dexterity, for unencumbered electrical PPE that remains easy to use
  • Suitable for wear over Class 1 to 4 electrical insulated gloves
Composed of premium goatskin leather, ActivArmr® 96-003 protectors guard against moderate cut, puncture and abrasion risks, while ensuring additional personal protection for workers engaged in electrical work, either with or near energized components.
Recommended for: Energized electrical, electrical, maintenance.
Certificaten: EN 388:2016 (2111B), ISO 9001, ASTM F496, ASTM F696
Verpakking: One pair per package.
Opgelet: Suitable for wear over Class 1 to 4 electrical insulated gloves.