EDGE® 48-707 Cut Resistant Gloves
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-707/10 48-707/11 48-707/6 48-707/7 48-707/8 48-707/9
ANSE48-707/10EA 112 EUR
ANSE48-707/10 ANSE48-707/11 ANSE48-707/6 ANSE48-707/7 ANSE48-707/8 ANSE48-707/9
EDGE® 48-707 Cut Resistant Gloves
Handschoenen
Medium-duty cut and abrasion-resistant gloves, offering high comfort and dexterity.
  • Seamless liner and knitted wrist: Added dexterity for meticulous work
  • Reinforced thumb crotch: Durable work gloves with abrasion resistance
  • FDA-approved food handling gloves: Superb safety and hygiene standards
  • Knitted from polyethylene, wrapped yarn, fiber glass, polyester, Spandex
Heightened grip and dexterity, owing to their palm PU coating, these protective gloves combine good grip as well as dexterity, and are suitable for dry or slightly oily environments.
Recommended for: Aeronautics, Assembly line, Assembling metal parts and components, Handling sharp or irregular metal pieces, Molding and Plastic injections.
Certificaten: EN 388:2016 (3X4XC), Cat. II
Food contact approved