Super-Tack™ Cleanroom Construction Tapes
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1112TB200-P3T 1112CL200-P3T 1112WH200-P3D
UTAP1112TB200-P3TEA 0 EUR
UTAP1112TB200-P3T UTAP1112CL200-P3T UTAP1112WH200-P3D
Super-Tack™ Cleanroom Construction Tapes
Tape
Ideal for any cleanroom construction.
  • Offer highest level of cleanliness available for today’s critical environments
  • Suitable for Class 1 (ISO class 3) to Class 1,000 (ISO Class 6) cleanrooms
Ultra tape's cleanroom manufacturing process ensures that unwanted contaminants are removed from both sides of the facestock.