Bizflame™ 88/12 FR95 Hi-Vis Shirts
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR95YERL FR95YERM FR95YERS FR95YERXL FR95YERXXL FR95YERXXXL
PRTWFR95YERLEA 152 EUR
PRTWFR95YERL PRTWFR95YERM PRTWFR95YERS PRTWFR95YERXL PRTWFR95YERXXL PRTWFR95YERXXXL
Bizflame™ 88/12 FR95 Hi-Vis Shirts
Werkkleding
This lightweight shirt offers guaranteed flame resistance for the life of the garment. Ideal for situations where extra visibility is required, FR95 is spacious and comfortable. Features include an action back for ease of movement and an extra long shirt tail to ensure the garment stays in place when tucked in. Shell Fabric: Bizflame 88/12 HV: 88% FR Cotton, 12% Nylon.
  • Dual hazard protection
  • Lightweight for enhanced wearer comfort
  • Action back for extra freedom of movement
  • Button cuffs for a secure and comfortable fit
  • Shirt tail hem stays tucked in
  • Protection against radiant, convective and contact heat
  • ARC2
  • Sew on flame resistant industrial wash tape
  • Two pockets for secure storage
  • 40+ UPF rated fabric to block 98% of UV rays
  • Available in sizes up to 6XL
Bizflame™ 88/12:
The fabric is soft yet hard wearing and the quality of the construction is excellent. The generous fit contributes to the overall appeal of this winning garment.
Flame Resistant:
This industry leading flame resistant range provides multi-standard protection for hazardous environments. These state of the art products are the result of years of experience combined with advanced technology and market research. Commitment to the health, safety and comfort of the wearer can be seen in the wide range of products suitable for all climates and end uses.
Length: 450 mm, Width: 370 mm, Height: 380 mm
Certificaten: ASTM F1959/F1959M-12 (ATPV 9 cal cm²(HAF 75,6%)), ASTM F1506, NFPA® 2112, NFPA® 70E, ANSI/ISEA 107 TYPE R CLASS 3, ANSI/ISEA 107 TYPE P CLASS 3, EN ISO 11612 (A1+A2, B1, C1, F1), EN ISO 20471 Class 3.