Automation Tips for PerkinElmer® (Revvity) Janus® MDT and Varispan Workstations
Leverancier: FLUOTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PKE-200.F PKE-200.NS PKE-200.ST PKE-200CB.F PKE-200CB.NS PKE-200CB.ST PKE-235.F PKE-235.NS PKE-235.ST PKE-30.F PKE-30.NS PKE-30.ST PKE-50.F PKE-50.NS PKE-50.ST PKE-50CB.F PKE-50CB.NS PKE-50CB.ST
736-1061EA 784 EUR
736-1061 736-1062 736-1063 736-1064 736-1065 736-1066 736-1067 736-1068 736-1069 736-1070 736-1071 736-1072 736-1073 736-1074 736-1075 736-1076 736-1077 736-1078
Automation Tips for PerkinElmer® (Revvity) Janus® MDT and Varispan Workstations
Pipettips
Fluotics manufactures a complete selection of automation tips compatible with the PerkinElmer® (Revvity) Janus® MDT and varispan workstations. These automation tips are directly interchangeable with their PerkinElmer® automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non-sterile, sterile, and barrier filter formats
Compatible with PerkinElmer® (Revvity) Janus® MDT and varispan workstations. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.
Verpakking: Pipette tips are packaged 96 or 384 per rack, 50 racks per case.