Printer Accessories for Models M511 and M211
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176448 176449 176450 176499 176501
BRDY176448EA 135.13 EUR
BRDY176448 BRDY176449 BRDY176450 BRDY176499 BRDY176501
Printer Accessories for Models M511 and M211
Printers Document Printers
Accessories for Brady Printer Models M511 and M211.
  • M511 hard case - transport and store your M511 label printer
  • M511 magnet - magnet attaches the M511 printer to panels, I-beams and other magnetic-receptive surfaces
  • M511 utility hook - hang printer easily on a cart, tool belt, rack or bucket organiser
  • M511 and M211 power brick - compatible with all mobile devices, 70-minute charge time
  • M511 and M211 AC adapter - to power and charge M511 and M211 printers with power supply voltage: 110 to 240 V
Hard case - Protects your printer from impacts and dust. Keeps printer, accessories and label cartridges organised and on hand. Durable hard case allows for easy mobility.
Magnet - Get hands-free operation by affixing to a metal surface. Attaches to back of printer with screws to ensure stability.
Utility hook - Attaches to back of printer with screws to ensure stability. Made from heavy-duty steel for added durability.
Power brick - A fully-charged 10000mAh Power Brick can charge your M511 or M211 printer up to three times. LED digital display shows the percentage of power remaining. Outputs include: USB-C, USB-A1/A2.
AC adapter - USB A-to-C dongle allows you to charge the printer using a USB port in a vehicle or on a laptop, plus can be used to connect the M511 to Brady workstation label creation software.
Bestelinformatie: Power brick (BRDY176499) has a rechargeable lithium-ion battery, must ship ground.