ActivArmr® 96-002 Goatskin Leather Gloves
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
96-002/10 96-002/11 96-002/12 96-002/7 96-002/8 96-002/9
ANSE96-002/10EA 411 EUR
ANSE96-002/10 ANSE96-002/11 ANSE96-002/12 ANSE96-002/7 ANSE96-002/8 ANSE96-002/9
ActivArmr® 96-002 Goatskin Leather Gloves
Handschoenen
Goatskin leather protectors, for enhanced low-voltage glove life and improved hand safety. Designed to be worn over Class 0 and 00 electrical safety gloves, these leather protectors can significantly extend wear life.
  • Extend the life of rubber insulated gloves while providing excellent hand protection
  • Provides cut, puncture, and abrasion protection
  • Manufactured to ASTM D120 and EN 60903 standards
  • Suitable to wear with Class 0 and 00 class gloves
ActivArmr® 96-002 protectors are made from premium goatskin leather, enhancing hand safety when conducting electrical work with (or close to) energized components, while offering moderate abrasion, puncture, and cut resistance. The leather formulation also offers excellent dexterity, for genuinely easy-to-use electrical PPE.
Certificaten: EN 388:2016 (2111B), ISO 9001, ASTM F496, ASTM F696
Verpakking: Gloves are individually packaged by pair.