HyFlex® 11-729 Industrial Gloves
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-729/6 11-729/7 11-729/8 11-729/9 11-729/10 11-729/11 11-729/12
ANSE11-729/6EA 145 EUR
ANSE11-729/6 ANSE11-729/7 ANSE11-729/8 ANSE11-729/9 ANSE11-729/10 ANSE11-729/11 ANSE11-729/12
HyFlex® 11-729 Industrial Gloves
Handschoenen
It is a touchscreen-friendly polyurethane gloves that offer heightened comfort and dexterit.
  • Carbon insert in index finger and thumb: Touchscreen compatibility
  • Spandex in liner and coating: For PPE gloves with ample dexterity
  • INTERCEPT Cut Resistance: Enhanced hand protection
  • Coating material: Polyurethane
Heightened tactility: Carbon inserts in the HyFlex® 11-729's index finger and thumb increase these industrial gloves tactile sensitivity, ensuring touchscreen compatibility.
Elevated comfort: HPPE Lycra® also present in the liner offers workers increased comfort by alleviating repetitive strain.
Enhanced dexterity: Spandex® in these industrial safety gloves coating and liner enables hand protection with increased dexterity and reduced hand fatigue.
Added protection: INTERCEPT Cut Resistance Technology’s blend of yarns and fibers ensures some cut protection.
Improved grip: HyFlex® 11-729 industrial working gloves polyurethane coating ensures good grip performance, for secure handling in dry or slightly oily conditions.
This is used in the industries like automobile, transportation, construction, glass manufacturing, machinery and equipment, maintenance, metal fabrication.
It is recommanded for aeronautics, assembling metal parts and components, assembly of white goods, glass repair and replacement, handling sharp or irregular metal pieces, press injection molding.
Certificaten: This is certified of EN ISO 21420:2020.