Zarate Trousers with ARC Protection
Leverancier: Sioen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Zarate
SIOE11VA2PFAB9856LEA 98.3 EUR
SIOE11VA2PFAB9856L SIOE11VA2PFAB9848R SIOE11VA2PFAB9848S SIOE11VA2PFAB9852L SIOE11VA2PFAB9848L SIOE11VA2PFAH4650S SIOE11VA2PFAH4650R SIOE11VA2PFAH4662R SIOE11VA2PFAH4654S SIOE11VA2PFAH4654R SIOE11VA2PFAB9844R SIOE11VA2PFAB9856S SIOE11VA2PFAH4650L SIOE11VA2PFAH4658S SIOE11VA2PFAH4646R SIOE11VA2PFAB9856R SIOE11VA2PFAH4658R SIOE11VA2PFAB9864R SIOE11VA2PFAM4446L SIOE11VA2PFAB9852S SIOE11VA2PFAH4654L SIOE11VA2PFAB9852R SIOE11VA2PFAB9860R SIOE11VA2PFAH4646L SIOE11VA2PFAM4454R SIOE11VA2PFAM4454S SIOE11VA2PFAM4450L SIOE11VA2PFAM4458R SIOE11VA2PFAM4446R SIOE11VA2PFAM4458S SIOE11VA2PFAM4454L SIOE11VA2PFAM4450R SIOE11VA2PFAM4450S SIOE11VA2PFAM4462R SIOE11VA2PFAB9846L SIOE11VA2PFAB9854L SIOE11VA2PFAB9858S SIOE11VA2PFAB9846R SIOE11VA2PFAB9858R SIOE11VA2PFAH4660R SIOE11VA2PFAH4652S SIOE11VA2PFAH4652R SIOE11VA2PFAH4664R SIOE11VA2PFAH4644R SIOE11VA2PFAH4656S SIOE11VA2PFAH4656R SIOE11VA2PFAM4456L SIOE11VA2PFAB9850L SIOE11VA2PFAH4652L SIOE11VA2PFAH4648S SIOE11VA2PFAB9854S SIOE11VA2PFAH4648R SIOE11VA2PFAB9854R SIOE11VA2PFAB9862R SIOE11VA2PFAM4448L SIOE11VA2PFAB9850S SIOE11VA2PFAH4656L SIOE11VA2PFAB9850R SIOE11VA2PFAH4648L SIOE11VA2PFAM4464R SIOE11VA2PFAM4456R SIOE11VA2PFAM4456S SIOE11VA2PFAM4444R SIOE11VA2PFAM4452L SIOE11VA2PFAM4448S SIOE11VA2PFAM4448R SIOE11VA2PFAM4460R SIOE11VA2PFAM4452R SIOE11VA2PFAM4452S
Zarate Trousers with ARC Protection
Werkkleding
Multifunctional and anti-static ARC trousers that provide excellent protection against arcs, heat, and metal splashes.
  • Closure by 1 hidden button/front fly with zip closure
  • 2 inset pockets with flap
  • 1 back pocket with flap closed by concealed press studs
  • 1 patched thigh pocket with flap closed by hidden press studs
  • 1 ruler pocket with flap closed by concealed press studs
  • Elasticated waist
  • Knee pockets
  • Waistband with belt loops
The trousers are made of a breathable and non-allergenic fabric and are very practical with two inset pockets with flap, one back pocket, one thigh pocket and one ruler pocket with concealed press stud fastening and reflective piping on the pocket flap. The trousers are provided with knee pockets where knee protection can be inserted.
Fabric: Sio-Safe™ Essential: 79% FR cotton+20% PES+1% AST; ±350 g/m²
Certificaten: IEC 61482-2:2009 class 1- ATPV 15.2 cal/cm²; EN ISO 11612:2015/A1A2B1C1E3F1; EN ISO 11611:2015/Class 1 - A1 A2; EN 1149-5:2008; EN 13034:2005+A1:2009 Type PB [6]; EN ISO 13688:2013; Tested for harmful substances according to Oeko-Tex® Standard 100 (1708040/Centexbel).