Manual Valves, CR4
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CR4-2M-4F-3 CR4-2M-4F
ENTECR4-2M-4F-3EA 0 EUR
ENTECR4-2M-4F-3 ENTECR4-2M-4F
Manual Valves, CR4
Ventielen Microventielen
Smaller than most comparable valves, ¹/₄" CR4 series valves are ideal for handling wet etch and clean process chemicals. These durable valves can handle temperatures up to 160 °C at 276 kPa.
  • Compact, modular design
  • High-temperature capable, all-PFA construction
  • High-purity corrosive chemical handling
  • All semiconductor WEC process chemicals
  • Chemical line sizes 3/8" or smaller
With no exposed metal hardware, the multi-turn valve is completely sealed and protected from harsh chemical environments. This all-PFA valve is available for high-purity chemical handling in line sizes 3/8" or smaller. A variety of connections including PrimeLock® , Flaretek® , FNPT, PureBond® , 'SpaceSaver', and Super 300 Type Pillar® allow flexibility in your fluid design. And, the compact, modular design enables manifold customisation.