SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator
Leverancier: THERMO SAVANT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SRF110P2-230 SRF110P1-230
THSASRF110P2-230EA 22900 EUR
THSASRF110P2-230 THSASRF110P1-230
SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator
Concentratoren
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules.
  • Powdered coated steel material
  • Self aligning and quick release rotor mechanism
  • Three preset program with option to store up to nine programs
  • Temperature range −4 to +100 °C
They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
Certificaten: UL, UKCA, KC, RCM certified.
Verpakking: SRF110P1 kit includes refrigerated SpeedVac (SRF110), hexagonal rotor (RF132), an oil pump (VLP120-115), a vapor Trap (RVT450-115), universal tubing (UTP-TYG), 4 L capacity glass condensation flask (GCF400), glass cap (FC400), and 1 L bottle of heat transfer fluid Cryocool™ (SCC1). SRF110P2 kit includes refrigerated SpeedVac (SRF110), hexagonal rotor (RF132), universal vacuum system (UVS450), universal tubing (UTP-TYG), 4 L capacity glass condensation flask (GCF400), glass cap (FC400), and 1 L bottle of heat transfer fluid Cryocool™ (SCC1).