AlphaTec® 58-005 Chemical Resistant Gloves
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
58-005/10 58-005/11 58-005/12 58-005/7 58-005/8 58-005/9
ANSE58-005/10EA 71.4 EUR
ANSE58-005/10 ANSE58-005/11 ANSE58-005/12 ANSE58-005/7 ANSE58-005/8 ANSE58-005/9
AlphaTec® 58-005 Chemical Resistant Gloves
Handschoenen
Mediumweight chemical-resistant gloves with excellent grip, dexterity and comfort. Silicone free.
  • Nylon liner integration: Enhanced dexterity and tactility
  • Ansell GRIP™ Technology: Safer handling in dry and wet conditions
  • Folded cuff gutter: For added safety and comfort levels
Elevated comfort: AlphaTec® 58-005 chemical-resistant gloves integrate a nylon liner into the nitrile and neoprene shell for better dexterity and tactility. Enhanced grip: These safety gloves feature Ansell GRIP™ Technology, enhancing dexterity and grip for secure handling of dry, wet or oily parts, as well as reducing hand fatigue.
Heightened protection: AlphaTec® 58-005 hand protection gloves offer a foldable cuff gutter, which prevents chemicals from running down the arm, providing added personal protection. Advanced features: AlphaTec® 58-005 chemical protection gloves are made of a unique nitrile and neoprene blend, ensuring enhanced chemical permeation and degradation resistance.
Recommended for: Assembly and inspection of components, oil, fluids and filter change, painting or cleaning tools or robots, handling incoming goods, transferring liquids and solids between vessels and tanks and process equipment, transferring liquids and solids, opening furnaces draining pumps valves or lines and crackers BTX process, cleaning furnaces, distillation pumps, valves or lines and crackers, BTX process, opening and draining pumps valves or lines.
Certificaten: EN ISO 374-1:2016 Type A (AJKLMNOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4122A), EN 407 (X1XXXX)
Verpakking: 6 pairs per bag.