Cleanroom Boots, with UG ESD Sole
Leverancier: ALSICO HIGHTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1028850-2XL 1028850-L 1028850-M 1028850-S 1028850-XL 1028850-XS
ALSO1028850-2XLEA 0 EUR
ALSO1028850-2XL ALSO1028850-L ALSO1028850-M ALSO1028850-S ALSO1028850-XL ALSO1028850-XS
Cleanroom Boots, with UG ESD Sole
Werkkleding
These boots are made of 100% nitrilic rubber with lightweight sole (±233 g).
  • Long durability
  • With ultra grip ESD sole and ESD logo
  • Best conductivity due to increased carbon
  • Safety profile: anti-slip
Features two coloured straps with 2-piece buckle on the foot. Closure with strap and studs on the upper side of the boot and reinforcement tape around the sole.
Certificaten: IEC 61340-5-1.