Automation Tips for Analytik Jena Cybio Felix, Well Vario, and Selma Workstations
Leverancier: FLUOTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CYBF-250.F CYBF-250.NS CYBF-250.ST CYBF-250TT.F CYBF-250TT.NS CYBF-250TT.ST CYBF-40Q.F CYBF-40Q.NS CYBF-40Q.ST CYBF-40TT.F CYBF-40TT.NS CYBF-40TT.ST CYBF-50.F CYBF-50.NS CYBF-50.ST CYBF-60Q.F CYBF-60Q.NS CYBF-60Q.ST CYBF-60TT.F CYBF-60TT.NS CYBF-60TT.ST CYBW-10.NS CYBW-10.ST CYBW-10Q.NS CYBW-10Q.ST CYBW-25.F CYBW-25.NS CYBW-25.ST CYBW-25Q.F CYBW-25Q.NS CYBW-25Q.ST CYBW-60.F CYBW-60.NS CYBW-60.ST CYBW-60Q.F CYBW-60Q.NS CYBW-60Q.ST
736-0964EA 784 EUR
736-0964 736-0965 736-0966 736-0967 736-0968 736-0969 736-0970 736-0971 736-0972 736-0973 736-0974 736-0975 736-0976 736-0977 736-0978 736-0979 736-0980 736-0981 736-0982 736-0983 736-0984 736-0985 736-0986 736-0987 736-0988 736-0989 736-0990 736-0991 736-0992 736-0993 736-0994 736-0995 736-0996 736-0997 736-0998 736-0999 736-1007
Automation Tips for Analytik Jena Cybio Felix, Well Vario, and Selma Workstations
Pipettips
Fluotics manufactures a selection of automation tips compatible with Analytik Jena Cybio Felix, Well Vario, and Selma Workstations. These automation tips are directly interchangeable with their Beckman automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non sterile, sterile, and barrier filter formats
Compatible with Analytik Jena Cybio Felix, Well Vario, and Selma Workstations. Available in quadrant and custom fill options. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.