BradyGrip® Print-on Hook Material for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170727 170728 170729
BRDY170727EA 35.51 EUR
BRDY170727 BRDY170728 BRDY170729
BradyGrip® Print-on Hook Material for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Easily place, remove and reposition labels on cable bundles with the BradyGrip® print-on hook label material (B-414), paired with VELCRO® ONE-WRAP® rolls. Available for Brady portable label printers.
  • Repositionable
  • Highly pliable
  • Fuel and oil resistant
  • Abrasion resistant
  • Matt finish
Constructed for long-term use and reuse. Designed with easy and safely mark wire and cable bundles. Easy to reposition and reuse when you need to update your bundles.
Matte finish, abrasion-resistant, humidity-resistant, temperature-resistant, indoor use B-414 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.