Polyester Pane Component Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170736 170730 170731 170732 170733 170734 170735 170737 170738 170739
BRDY170736EA 54.47 EUR
BRDY170736 BRDY170730 BRDY170731 BRDY170732 BRDY170733 BRDY170734 BRDY170735 BRDY170737 BRDY170738 BRDY170739
Polyester Pane Component Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady handheld label printer.
  • High adhesion
  • Abrasion resistant
  • Fuel and oil resistant
  • Heat resistant
  • Low temperature resistant
  • Gloss finish
Designed for a wide range of applications such as labelling equipment, circuit boards, panel components and barcodes. Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water. Made with a permanent acrylic adhesive that sticks toMore Product Information