Module for Remote Signalling
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38765 39221
MSGA38765EA 384 EUR
MSGA38765 MSGA39221
Module for Remote Signalling
Meubilair Opslagkasten
The module offers an alarm forwarding via the mobile phone network (SMS/call) and is therefore especially suitable for companies and facilities without a central building management system. In an emergency, the immediate alerting of one or several defined persons is triggered.
  • The module is easily installable on-site via plug
  • Five programmable telephone numbers
  • Configurable message texts
  • Collective alarm in case of an emergency or power failure
A SIM card is required for the initial operation of the module, which must be provided by the customer.More Product Information