Minigrip® Bags, Zip Seal, Printed Kangaroo Pouch
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM020711 ITM020712 ITM020713
129-3873EA 255 EUR
129-3873 129-3874 129-3875
Minigrip® Bags, Zip Seal, Printed Kangaroo Pouch
Zakken Monsterzakken
Clear PE bags, thickness 60 µm, food contact.
  • Extra external pocket to protect accompanying documents
  • Suitable for food contact
  • Hanging hole
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Bags printed with the reference 'NON STERILE CUSTOM MEDICAL DEVICE' according to the EU directive 93/42/CEE. This reference is printed in 4 languages (French, English, Spanish and German).
Certificaten: Suitable for food contact. Certificate available on request.