PermaSleeve® Wiremarker Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Label Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170718 170712 170713 170714 170715 170716 170717 170719 170720 170721 170722
BRDY170718EA 69.09 EUR
BRDY170718 BRDY170712 BRDY170713 BRDY170714 BRDY170715 BRDY170716 BRDY170717 BRDY170719 BRDY170720 BRDY170721 BRDY170722
PermaSleeve® Wiremarker Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Label Printers
Etiketten
Permanently mark wires with the durable PermaSleeve® PS heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Brady handheld label printer.
  • Specialised - Designed specifically for rigorous cable and wire marking applications
  • Durable - Self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel and oil
  • Efficient - Fast shrinking to save installation time and cost
  • Easily Identifiable - Made with a crisp, clear identification surface that’s resistant to abrasions and fading
Applications include electrical labelling and wire and cable labelling.
Matte finish, chemical-resistant, corrosion-resistant, humidity-resistant, smear-resistant, UV-resistant, weather-resistant B-342 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certificaten: UL Recognised to UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1), SAE AS-81531, MIL-STD-202.