High Visibility Windbreaker Fleece Jackets, F303
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F303YERL F303YERM F303YERS F303YERXL F303YERXXL F303YERXXXL
PRTWF303YERLEA 48.1 EUR
PRTWF303YERL PRTWF303YERM PRTWF303YERS PRTWF303YERXL PRTWF303YERXXL PRTWF303YERXXXL
High Visibility Windbreaker Fleece Jackets, F303
Werkkleding
The Hi-Vis windbreaker fleece offers great wearer warmth and comfort. With an added layer to keep the wind out.
  • Anti-pill durable fleece fabric
  • Reflective tape for increased visibility
  • Elastic waist with toggle for a secure and comfortable fit
  • Elasticated cuffs for a secure fit
  • Unlined
  • 40 UPF rated fabric to block 98% of UV rays
  • Waterproof/breathability WP 8,000 mm, MVP 3,000g/m²/24 h
Features include two secure side pockets with zips, elasticated cuffs and an elasticated draw cord waist for a more comfortable fit. The high quality anti-pill finish maintains the look of the garment for longer.
Certificaten: EN ISO 20471 Class 2 for size S and EN ISO 20471 Class 3 for size M upwards.