Jugs, Tapered, Stainless Steel
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PASJ-TJ06 PASJ-TJ1 PASJ-TJ3 PASJ-TJ5 PASJ-TJ2
TEKMPASJ-TJ06EA 0 EUR
TEKMPASJ-TJ06 TEKMPASJ-TJ1 TEKMPASJ-TJ3 TEKMPASJ-TJ5 TEKMPASJ-TJ2
Jugs, Tapered, Stainless Steel
Flessen Voorraadflessen
Tapered jugs are available in a number of sizes upto five litres, and are designed for hygiene, easy cleaning and durability.
  • Fully welded 304-grade stainless steel
  • Bright polished finish
  • Crevice free