Minigrip® Antistatic Zip Seal Bags
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM025045 ITM025046 ITM025047 ITM025048 ITM025049 ITM025050 ITM025054 ITM025052 ITM025053
129-3878EA 114 EUR
129-3878 129-3879 129-3880 129-3881 129-3882 129-3883 129-3884 129-3885 129-3886
Minigrip® Antistatic Zip Seal Bags
Zakken Monsterzakken
Minigrip® antistatic zip seal bags made up of PE.
  • Average surface resistivity of ±1012 Ohms
  • Half decay time of ±3 s
  • Gives protection to the contents inside