Indoor Outdoor Vinyl Labels for M410, BMP41, M510, BMP51 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176803 176804 176805 176806 176807
BRDY176803EA 47.59 EUR
BRDY176803 BRDY176804 BRDY176805 BRDY176806 BRDY176807
Indoor Outdoor Vinyl Labels for M410, BMP41, M510, BMP51 and M511 Printers
Etiketten
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.
  • Outdoor durable
  • Low temperature resistant
  • Fuel and oil resistant
  • High adhesion
  • Abrasion resistant
Versatile - Ideal for labeling pipes, equipment, shelves and other industrial surfaces
Tough - Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
Adhesive - Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
Long-lasting - Weather-resistant labels designed to last up to 10 years in an outdoor environment
Use this versatile outdoor-friendly material when multiple color options will clearly provide information when adhered to rugged surfaces in dirty conditions.
Common applications: Indoor/outdoor, safety/facility, equipment ID, general ID, DataCom
Ribbon series: R6000HF
Thickness: 0,102 mm
Environment: Freezer
Min. application temperature: −18 °C
Max. service temperature: 82 °C
Min. service temperature: −40 °C
Gloss finish, abrasion-resistant, chemical-resistant, oil-resistant, water-resistant, outdoor use B-595 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, and M511.
Certificaten: Compliant with Directive 2011/65/EU and 2015/863, BS 5609:1986 Section 2 and section 3.