Aggressive Adhesive Multi-Purpose Clear Polyester Labels B-432 Tape for M6, M7 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174182 173910 173880 173728 173685 173693 173878
BRDY174182EA 145.2 EUR
BRDY174182 BRDY173910 BRDY173880 BRDY173728 BRDY173685 BRDY173693 BRDY173878
Aggressive Adhesive Multi-Purpose Clear Polyester Labels B-432 Tape for M6, M7 Printers
Etiketten
Rough surface polyester (B-432) labels are ideal for electronic component identification, barcode labels, rating plates and solar panel identification.
  • Made from durable glossy clear polyester with permanent acrylic adhesive, labels are chemical, oil and water resistant
  • UL recognised for outdoor use on glass, thermoset polyester plastic and polyvinyl fluoride plastic surfaces to support solar panel identification
  • CSA accepted
  • Service temperature of −56 to +110 °C
  • Shape: Rectangle
Brady materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Application: Asset tracking, barcode labeling, component and equipment labeling, data and telecommunications labeling, electrical labeling, general identification, inventory and inspection labeling, warehouse marking.
Resistance properties: Chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant.