rubis® Tweezers, Reverse Action, Anti-Wicking, Stainless Steel
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C44AWG28-30 C45AWG24-26 C46AWG20-22 C47AWG16-18
OUTIC44AWG28-30EA 52.7 EUR
OUTIC44AWG28-30 OUTIC45AWG24-26 OUTIC46AWG20-22 OUTIC47AWG16-18
rubis® Tweezers, Reverse Action, Anti-Wicking, Stainless Steel
Pincetten
Tweezers, reverse action, anti-wicking, antimagnetic. Made up of stainless steel.
  • Comfortable soldering tweezers
  • Insertion/extraction tweezers
  • Eliminates wicking, protects plastic insulation on wire and acts as a heat sink while soldering
  • Point style: Round