VOID Pattern Tamper Evident Matte Metallised Polyester Labels for M6 M7 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173871 173733 173830 173747
BRDY173871EA 77.12 EUR
BRDY173871 BRDY173733 BRDY173830 BRDY173747
VOID Pattern Tamper Evident Matte Metallised Polyester Labels for M6 M7 Printers
Etiketten
The B-7576 material is designed to leave a 'VOID' footprint when the label is removed, the footprint also appears on the surface of the label so it cannot be reused.
  • Ideal for use in rating and serial plates that require high performance and evidence of label removal
  • Use care when handling to maintain the VOID pattern
  • For use in thermal transfer printers
  • Shape: Rectangle
  • Service temperature range: −70 to −135,5 °C
Brady materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Resistance properties: Chemical-resistant, humidity-resistant, UV-resistant, weather-resistant.
Application: General identification, inventory and inspection labeling, security identification.
Certificaten: RoHS compliant to Pass 2011/65/EU.