Matte White 2 mil Polyimide Circuit Board Labels, B33
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153217 153219 153213 153216 153209 153214 153211 153210 153212 153220 153215
BRDY153217EA 1540.91 EUR
BRDY153217 BRDY153219 BRDY153213 BRDY153216 BRDY153209 BRDY153214 BRDY153211 BRDY153210 BRDY153212 BRDY153220 BRDY153215
Matte White 2 mil Polyimide Circuit Board Labels, B33
Etiketten
Permanent Polyimide (B-728) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labeling.
  • Matte white polyimide material is thermal transfer printable
  • Service temperature of −70 to 100 °C
  • A ribbon is required to print this material
Ultra-durable adhesive is designed for use with extreme wash protocol and cleaning chemicals, and is ideal for use in auto apply equipment.
Ribbon series: B30 monocolour series
Shape: Rectangle
Label properties: Heat resistant, permanent adhesive
Resistance properties: Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant
Applications: Circuit board labelling, component and equipment labelling.
Certificaten: UL approved; Meets requirements of MIL-STD-202G, Method 215K.