Metalised Polyester Labels for M610 and M710 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174231 173869 173803 173873 173816 173827 173717 173840 173729
BRDY174231EA 39.43 EUR
BRDY174231 BRDY173869 BRDY173803 BRDY173873 BRDY173816 BRDY173827 BRDY173717 BRDY173840 BRDY173729
Metalised Polyester Labels for M610 and M710 Printers
Etiketten
Glossy metallised polyester labels (B-435A) are designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
  • High Adhesion - Permanent acrylic adhesive withstands numerous solvents and variable temperatures
  • Service temperature of –40 to 90 °C
  • Label Properties - Chemical resistant, metallic, permanent adhesive
Application: Asset tracking, circuit board labeling, component and equipment labeling, general identification, inventory and inspection labeling. Resistance properties: Abrasion-resistant, chemical-resistant, humidity-resistant, UV-resistant, weather-resistant.