VOID Pattern Tamper Evident Polyester Labels for M610 and M710 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173732 173829 173746
BRDY173732EA 83.41 EUR
BRDY173732 BRDY173829 BRDY173746
VOID Pattern Tamper Evident Polyester Labels for M610 and M710 Printers
Etiketten
Tamper-Evident Polyester (B-7546) leaves a 'VOID' footprint when the label is removed and leaves a 'VOID' pattern on the top surface of the label to prevent reuse.
  • Gloss white polyester with tamper indicating acrylic is ideal for rating and serial plates that require high performance and evidence of label removal
  • Excellent chemical, oil and water resistance
  • Service temperature of –20 to 100 °C
Label properties: Heat resistant, permanent adhesive, tamper evident. Resistance properties: chemical-resistant, humidity-resistant, UV-resistant, weather-resistant.
Application: General identification, inventory and inspection labeling, security identification.
Certificaten: UL Recognised; CSA accepted.