Cerex® SPE Columns HPWCX
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30174772 30215446
TCAT30174772EA 0 EUR
TCAT30174772 TCAT30215446
Cerex® SPE Columns HPWCX
Kolommen voor chromatografie
Polymer monodisperse weak cation exchange chemistry, used for retention of strong bases in biological fluids. Ideal for strong base extraction for drug monitoring, screening, identification, confirmation, and quantitation.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible