Cerex® WWP SPE Columns
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
676-0201R 676-0353C
TCAT676-0201REA 0 EUR
TCAT676-0201R TCAT676-0353C
Cerex® WWP SPE Columns
Kolommen voor chromatografie
Microparticulate water wettable polymer provides great chemical stability and ideal for synthetic cannabinoids analysis.