Cerex® OFX SPE Columns
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
633-0101R 633-0353C
TCAT633-0101REA 0 EUR
TCAT633-0101R TCAT633-0353C
Cerex® OFX SPE Columns
Kolommen voor chromatografie
Proprietary blend of ion-exchange and water-wettable polymer targets towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.